missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-octylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals

Conducting polymers

$402.67 - $1326.14

Chemical Identifiers

CAS 104934-51-2
Molecular Formula (C12H18S)n
Molecular Weight (g/mol) 241.44
MDL Number MFCD00241412
InChI Key RXIQSINDOTYIGV-UHFFFAOYSA-N
Synonym poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
PubChem CID 121233643
IUPAC Name azane;2,5-dimethyl-3-octylthiophene
SMILES CCCCCCCCC1=C(-*)SC(-*)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4477701
View Documents
Thermo Scientific Chemicals
04477701
0.25 g
Each for $402.67
Only null left
 
AA4477703
View Documents
Thermo Scientific Chemicals
04477703
1 g
Each for $1,326.14
Only null left
 
Description

Description

It is employed Conducting polymer, rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials. It is also used in organic field-effect transistors and in polymer-based solar cells and p-type polymer semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

104934-51-2
241.44
RXIQSINDOTYIGV-UHFFFAOYSA-N
121233643
CCCCCCCCC1=C(-*)SC(-*)=C1
(C12H18S)n
MFCD00241412
poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
azane;2,5-dimethyl-3-octylthiophene

Specifications

Powder
(C12H18S)n
poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
0.25 g
CCCCCCCCC1=C(-*)SC(-*)=C1
241.44
Poly (3-octylthiophene-2,5-diyl), Regioregular, low metals
104934-51-2
MFCD00241412
Soluble in THF,CHCl3
RXIQSINDOTYIGV-UHFFFAOYSA-N
azane;2,5-dimethyl-3-octylthiophene
121233643
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only