missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pleiotrophin Monoclonal Antibody (K06279_4G7), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA555682

 View more versions of this product

Catalog No. MA555682


Only null left
Explore available promotions

Description

Description

Pleiotrophin (PTN) is an 18 kDa secreted heparin and chondroitin-binding protein that is a regulatory cytokine critical to the early development of blood vessels and neurons. High PTN serum levels are associated with a variety of solid tumors. It was shown that patients with multiple myeloma (MM) also have elevated serum levels of this protein. The expression of this protein is very limited in normal adult tissues.
Specifications

Specifications

Pleiotrophin
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P21246, P63089
PTN
Recombinant human PTN (proprietary range information).
50 μg
Primary
Human, Mouse
Antibody
IgG1
Western Blot
K06279_4G7
Unconjugated
PTN
HARP; HBBM; HBBN; HB-GAM; HBGF8; HBGF-8; HBNF; HBNF1; HBNF-1; heparin affin regulatory peptide; heparin affin regulatory protein; heparin binding factor; heparin-binding brain mitogen; Heparin-binding growth factor 8; heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; heparin-binding neurite promoting factor; heparin-binding neurotorphic factor; heparin-binding neutrophic factor; NEGF1; OSF; Osf1; OSF-1; osteoblastic cell factor; Osteoblast-specific factor 1; pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); Pleiotrophin (Heparine binding factor, Hbnf, in the mouse); PTN
Mouse
Protein A
RUO
19242, 5764
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions