missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKD2 Monoclonal Antibody (K49022_2E5), Invitrogen™
Mouse Monoclonal Antibody
$1557.73 - $2745.56
Specifications
Antigen | PKD2 |
---|---|
Clone | K49022_2E5 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Description
Sequence of this protein is as follows: SDLIRKGYHK ALVKLKLKKN TVDDISESLR QGGGKLNFDE LRQDLKGKGH TDAEIEAIFT KYDQDGDQEL TEHEHQQMRD DLEKEREDLD LDHSSLPRPM SSRSFPRSLD DSEEDDDEDS GHSSRRRGSI SSGVSYEEFQ VLVRRVDRME HSIGSIVSKI DAVIVKLEIM ERAKLKRREV LGRLLDGVAE DERLGRDSEI HREQMERLVR EELERWESDD AASQISHGLG TPVGLNGQPR PRSSRPSSSQ
PKD2 Monoclonal antibody specifically detects PKD2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
PKD2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide | |
A MGC138466; APKD2; Autosomal dominant polycystic kidney disease type II protein; C030034P18Rik; MGC138468; PC2; Pc-2; PKD2; PKD4; polycystic kidney disease 2; polycystic kidney disease 2 (autosomal dominant); polycystic kidney disease 2 homolog; Polycystic kidney disease 2 protein; Polycystic kidney disease 2 protein homolog; polycystin 2; polycystin 2, transient receptor potential cation channel; polycystin-2; Polycystwin; R48321; RGD1559992; transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2; TRPP1; TRPP2 | |
Recombinant human PKD2 (immunogen range is 701-950). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
PKD2 |
K49022_2E5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O35245, Q13563 | |
18764, 5311 | |
IgG1 | |
Protein A | |
Antibody |