Learn More
Pipette Supplies Inc PIPETMAN TIP EJECTOR P100 DFS Item
Supplier:  Pipette Supplies Inc GIF144605
Tip ejectors for:Gilson Pipetman Classic 100L (P100)Gilson Pipetman Fixed F25, F50, F100Gilson Pipetman Neo 100L (P100N)Made in France
Catalog No. 501317632
Only null left
Explore available promotions