missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine Dihydrochloride 98.0+%, TCI America™
$124.73 - $326.94
Chemical Identifiers
CAS | 142-64-3 |
---|---|
Molecular Formula | C4H12Cl2N2 |
Molecular Weight (g/mol) | 159.054 |
MDL Number | MFCD00012754 |
InChI Key | CVVIJWRCGSYCMB-UHFFFAOYSA-N |
Synonym | piperazine dihydrochloride, dowzene dhc, piperazine, dihydrochloride, dihydro pip wormer, caswell no. 668, piperazine wormer premix, unii-17vu4z4w88, diethylenediamine dihydrochloride, piperazine hcl, epa pesticide chemical code 067401 |
PubChem CID | 8893 |
IUPAC Name | piperazine;dihydrochloride |
SMILES | C1CNCCN1.Cl.Cl |
Chemical Identifiers
142-64-3 | |
159.054 | |
CVVIJWRCGSYCMB-UHFFFAOYSA-N | |
8893 | |
C1CNCCN1.Cl.Cl |
C4H12Cl2N2 | |
MFCD00012754 | |
piperazine dihydrochloride, dowzene dhc, piperazine, dihydrochloride, dihydro pip wormer, caswell no. 668, piperazine wormer premix, unii-17vu4z4w88, diethylenediamine dihydrochloride, piperazine hcl, epa pesticide chemical code 067401 | |
piperazine;dihydrochloride |
Specifications
142-64-3 | |
White | |
MFCD00012754 | |
piperazine dihydrochloride, dowzene dhc, piperazine, dihydrochloride, dihydro pip wormer, caswell no. 668, piperazine wormer premix, unii-17vu4z4w88, diethylenediamine dihydrochloride, piperazine hcl, epa pesticide chemical code 067401 | |
C1CNCCN1.Cl.Cl | |
159.054 | |
159.05 | |
Crystalline Powder |
320°C | |
C4H12Cl2N2 | |
1 g | |
CVVIJWRCGSYCMB-UHFFFAOYSA-N | |
piperazine;dihydrochloride | |
8893 | |
≥98.0% (N,T) | |
Piperazine Dihydrochloride |
Safety and Handling
RTECSNumber : TL4025000
TSCA : Yes