missing translation for 'onlineSavingsMsg'
Learn More

Piperazine, Anhydrous, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor PI12325KGBL

 View more versions of this product

Catalog No. 18-612-384


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

110-85-0
108° to 113°C
C4H10N2
GLUUGHFHXGJENI-UHFFFAOYSA-N
piperazine
0.99
1
Fiber Drum
25 kg
C1CNCCN1
86.14
Reagent

Chemical Identifiers

110-85-0
86.14
piperazine
C4H10N2
GLUUGHFHXGJENI-UHFFFAOYSA-N
C1CNCCN1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.