missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid, Crystal, Reagent, ACS, Spectrum™ Chemical

P1145, 88-89-1, C6H2(NO2)3OH

Chemical Identifiers

CAS 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) 229.10
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
IUPAC Name 2,4,6-trinitrophenol
SMILES OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605212
View Documents
Spectrum Chemical Mfg Cor
P114525GM
25 g Amber Glass Bottle N/A
Only null left
 
Description

Description

Spectrum™ Chemical Picric Acid, Crystal, Reagent, ACS , also known as 2,4,6-trinitrophenol or TNP. It is a yellow crystalline solid and one of the most acidic phenols. As a highly nitrated compound, picric acid is highly explosive (like TNT). Apart from it's use as an explosive, it also has some uses in organic chemistry for the preparation of crystalline salts of organic bases. It is also used in clinical chemistry for the Jaffe reaction (a colorimetric method to determine creatinine levels in blood and urine).

Specifications

Chemical Identifiers

88-89-1
229.10
2,4,6-trinitrophenol
C6H3N3O7
OXNIZHLAWKMVMX-UHFFFAOYSA-N
OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

Specifications

88-89-1
C6H3N3O7
OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
229.10
25 g
1
OXNIZHLAWKMVMX-UHFFFAOYSA-N
2,4,6-trinitrophenol
ACS
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'