missing translation for 'onlineSavingsMsg'
Learn More

Phthalocyanine Chloroaluminum 98.0+%, TCI America™

$120.95 - $120.95

Chemical Identifiers

CAS 14154-42-8
Molecular Formula C32H16AlClN8
MDL Number MFCD00049386
Synonym Chloroaluminum Phthalocyanine
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C11671G
View Documents
TCI America
C11671G
1 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

14154-42-8
C32H16AlClN8
MFCD00049386
Chloroaluminum Phthalocyanine

Specifications

14154-42-8
C32H16AlClN8
1 g
574.97
Crystalline Powder
Blue-Purple
MFCD00049386
Chloroaluminum Phthalocyanine
≥98.0% (T)
Phthalocyanine Chloroaluminum
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'