missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide, 98%, Spectrum™ Chemical

$292.47 - $405.30

Chemical Identifiers

CAS 85-41-6
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
InChI Key XKJCHHZQLQNZHY-UHFFFAOYSA-N
IUPAC Name 2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1NC(=O)C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18-612-369
View Documents
Spectrum Chemical Mfg Cor
P1141500GM
500 g Poly Bottle
Each for $292.47
Only null left
 
18-612-368
View Documents
Spectrum Chemical Mfg Cor
P11411KG
1 kg Poly Pail
Each for $405.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

85-41-6
147.13
2,3-dihydro-1H-isoindole-1,3-dione
C8H5NO2
XKJCHHZQLQNZHY-UHFFFAOYSA-N
O=C1NC(=O)C2=CC=CC=C12

Specifications

85-41-6
231° to 236°C
C8H5NO2
XKJCHHZQLQNZHY-UHFFFAOYSA-N
2,3-dihydro-1H-isoindole-1,3-dione
0.98
1
Poly Bottle
500 g
O=C1NC(=O)C2=CC=CC=C12
147.13
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.