missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic Acid 99.0+%, TCI America™
$39.14 - $108.11
Chemical Identifiers
CAS | 88-99-3 |
---|---|
Molecular Formula | C8H6O4 |
Molecular Weight (g/mol) | 166.132 |
MDL Number | MFCD00002467 |
InChI Key | XNGIFLGASWRNHJ-UHFFFAOYSA-N |
Synonym | 1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid |
PubChem CID | 1017 |
ChEBI | CHEBI:29069 |
IUPAC Name | phthalic acid |
SMILES | C1=CC=C(C(=C1)C(=O)O)C(=O)O |
Chemical Identifiers
88-99-3 | |
166.132 | |
XNGIFLGASWRNHJ-UHFFFAOYSA-N | |
1017 | |
phthalic acid |
C8H6O4 | |
MFCD00002467 | |
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid | |
CHEBI:29069 | |
C1=CC=C(C(=C1)C(=O)O)C(=O)O |
Specifications
210°C | |
White | |
MFCD00002467 | |
XNGIFLGASWRNHJ-UHFFFAOYSA-N | |
phthalic acid | |
1017 | |
166.13 | |
25 g | |
Phthalic Acid |
88-99-3 | |
C8H6O4 | |
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid | |
C1=CC=C(C(=C1)C(=O)O)C(=O)O | |
166.132 | |
CHEBI:29069 | |
≥99.0% (GC,T) | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1337
RTECSNumber : TH9625000
TSCA : Yes