missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, FCC, 85%, Spectrum™ Chemical

P1097, 7664-38-2, H3PO4

Supplier:  Spectrum Chemical Mfg Cor P109720LTBL

 View more versions of this product

Catalog No. 18605175


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Phosphoric Acid, 85 Percent, FCC is used to acidify foods and beverages such as various colas and jams. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7664-38-2
Poly Drum
H3O4P
OP(O)(O)=O
97.99
FCC
1
20 L
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
85%

Chemical Identifiers

7664-38-2
97.99
phosphoric acid
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
OP(O)(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions