missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, FCC, 85%, Spectrum™ Chemical
P1097, 7664-38-2, H3PO4
Supplier: Spectrum Chemical Mfg Cor P10971LT
Description
Spectrum™ Chemical Phosphoric Acid, 85 Percent, FCC is used to acidify foods and beverages such as various colas and jams. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
7664-38-2 | |
Amber Glass Bottle | |
H3O4P | |
OP(O)(O)=O | |
97.99 | |
FCC |
1 | |
1 L | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
85% |
Chemical Identifiers
7664-38-2 | |
97.99 | |
phosphoric acid |
H3O4P | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
OP(O)(O)=O |