missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Salicylate, Crystal, Purified, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor PH16012KG
Specifications
118-55-8 | |
40° to 47°C | |
12 kg | |
Purified |
1 | |
Poly Pail | |
0.98 |
Chemical Identifiers
118-55-8 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.