missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenoxyethanol, Reagent, 99%, Spectrum™ Chemical
P1077, 122-99-6, C8H10O2
Supplier: Spectrum Chemical Mfg Cor P10774LTGL
Description
Spectrum™ Chemical Phenoxyethanol, Reagent , is a glycol ether and an organic chemical compound. This colorless oily liquid is often used in dermalogical products and often replaces sodium azide as a biological buffer. It can also be found as a preservative in cosmetics, vaccines and various pharmaceuticals. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Specifications
122-99-6 | |
244 to 250°C | |
C8H10O2 | |
QCDWFXQBSFUVSP-UHFFFAOYSA-N | |
2-phenoxyethan-1-ol | |
99% | |
Liquid |
1 | |
Amber Glass Bottle | |
4 L | |
OCCOC1=CC=CC=C1 | |
138.17 | |
Reagent |
Chemical Identifiers
122-99-6 | |
138.17 | |
2-phenoxyethan-1-ol |
C8H10O2 | |
QCDWFXQBSFUVSP-UHFFFAOYSA-N | |
OCCOC1=CC=CC=C1 |