missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenanthrene, 90%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor PH10625KG
Specifications
85-01-8 | |
Poly Pail | |
0.9 |
1 | |
2.5 kg | |
Reagent |
Chemical Identifiers
85-01-8 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.