Learn More
Haztech Systems Inc Hazcat pH Test Strips. Scale 0-14. DFS Item
Supplier:  Haztech Systems Inc HW4048
Hazcat pH Test Paper. These papers do not bleed, and can withstand the heat that builds inside the test tube when conducting a Thermal Analysis of a substance. Scale 0-14.
Catalog No. NC9259230
Only null left
Explore available promotions