missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite PetroPad
Reusable, washable and easy to dispose absorbent pad designed for the safe handling and the disposal of hazardous hydrocarbon waste.
$229.86 - $1712.59
Specifications
Material | Polymer |
---|---|
Type | Sorbent, Oil-only |
Disposable | Reusable |
Weight Capacity | Heavyweight |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
17-988-513
|
Justrite
83988 |
53.9 x 53.9 in. |
Each for $1,286.92
|
|
|||||
17-988-512
|
Justrite
83990 |
85.8 x 53.9 in. |
Each for $1,712.59
|
|
|||||
17-988-514
|
Justrite
83986 |
53.9 x 35.8 in. |
Each for $878.47
|
|
|||||
17-988-516
|
Justrite
83982 |
24 x 18.1 in. |
Each for $229.86
|
|
|||||
17-988-515
|
Justrite
83984 |
35.4 x 27.2 in. |
Each for $477.84
|
|
|||||
Description
- Portable and easy to fold up and store without fear of leakage or mess
- Absorbs and permanently solidifies hydrocarbons while allowing rainwater to pass through
- Durable material and robust construction withstand the demands of a rigorous work environment
- Easily pressure wash built-up dirt and debris without releasing hydrocarbons
- TCLP leachate test and EPA Paint Filter Test (9095B) compliant
Specifications
Polymer | |
Sorbent, Oil-only | |
Reusable | |
Heavyweight |