missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0% (C5 & C6 Isomers), Honeywell™ Burdick & Jackson
$2874.80 - $2874.80
Chemical Identifiers
CAS | 8032-32-4 |
---|---|
Synonym | p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Chemical Identifiers
8032-32-4 | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Specifications
8032-32-4 | |
0.64 g/cm3 | |
19 L | |
Metal Can | |
0.02% max. | |
>99% | |
Petroleum Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0% (C5 & C6 Isomers) |
100.0000% | |
35°C to 60°C | |
−18°C | |
UN1268 | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide | |
ACS Reagent | |
by GC total C5 and C6 isomers |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Petroleum distillates, n.o.s.