missing translation for 'onlineSavingsMsg'
Learn More

Sartorius PES Tangential Flow Filtration Cassette for up to 0.5 L

Plug-and-play tangential flow filtration cassette that is made from PES, for ultrafiltration and diafiltration of intermediate sample volumes.

Supplier:  Sartorius VFS050P0050IV

 View more versions of this product

Catalog No. 10-569-109


Only null left
Explore available promotions

Description

Description

  • Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
  • All-in-one design avoids the need for expensive cassette holders and TFF systems.
  • Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
  • High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
  • Improve safety for you and your sample with no need for flushing or cleaning.
  • Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
  • Membrane material: PES
  • Recommended feed volume: 100 to 500 mL
Specifications

Specifications

Ultrafiltration
Peristaltic Pump
PES
0.1 to 0.5 L
TFF cassette
Cassette
Polycarbonate
300 kDa
Non-sterile
0.5 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions