missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, Reagent Grade, Reagents

Periodic Acid, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$379.20 - $1214.00

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym Iodine Tetraoxide
IUPAC Name tetrahydroxyiodous acid
SMILES O[I](O)(O)(O)(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-337-879
View Documents
Labchem Inc
C1267250100C2
100 g
Each for $1,214.00
Only null left
 
01-337-878
View Documents
Labchem Inc
C126725025C1
25 g
Each for $379.20
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10450-60-9
227.94
Iodine Tetraoxide
O[I](O)(O)(O)(O)=O
H5IO6
TWLXDPFBEPBAQB-UHFFFAOYSA-N
tetrahydroxyiodous acid

Specifications

White to Pale Yellow
140°C
1.4
H5IO6
Iodine Tetraoxide
O[I](O)(O)(O)(O)=O
227.94
Periodic Acid, Reagent Grade
10450-60-9
Glass Bottle
100 g
Solid
TWLXDPFBEPBAQB-UHFFFAOYSA-N
tetrahydroxyiodous acid
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'