missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, 2.00 Normal, Aqueous, Reagents
Perchloric Acid, 2.00 Normal, Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
$159.13 - $227.33
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Chlorine Tetraoxide |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-334-290
|
Labchem Inc
CP1213001C |
1 L |
N/A
|
|
|||||
01-334-289
|
Labchem Inc
CP121300500C |
500 mL |
N/A
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
Chlorine Tetraoxide | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless | |
101°C | |
1.1145 | |
H2O | |
Chlorine Tetraoxide | |
O | |
18.02 | |
Perchloric Acid, 2.00 Normal, Aqueous |
7732-18-5,7601-90-3 | |
Glass Bottle | |
1 L | |
Liquid | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent |