missing translation for 'onlineSavingsMsg'
Learn More

Pentane CHROMASOLV™, for HPLC, ≥99.0%, Honeywell Riedel-de Haën™

CHROMASOLV™, for HPLC,≥99.0%

$253.13 - $3453.28

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
View More Specs

Alert:

Repeated exposure may cause skin dryness or cracking.


Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6001114
View Documents
Honeywell-Chromasolv
349561L
1 L
Each for $253.13
Only null left
 
6001115
View Documents
Honeywell-Chromasolv
3495625L
2.5 L
Each for $391.55
Only null left
 
6001117
View Documents
Honeywell-Chromasolv
349566X1L
6 x 1 L
Case of 6 Each for $1,230.55
Only null left
 
6001116
View Documents
Honeywell-Chromasolv
349564X25L
4 x 2.5 L
Case of 4 Each for $1,213.38
Only null left
 
60048243
View Documents
Honeywell Chemicals
349564X4L
4 x 4 L
Case of 4 Each for $3,453.28
Only null left
 
Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
CHEBI:37830
CCCCC
C5H12
MFCD00009498
8003
pentane

Specifications

-130°C
0.626g/mL at 25°C
1 L
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CCCCC
72.15
CHEBI:37830
≥99%
Pentane, for HPLC
109-66-0
35°C to 36°C
n20/D 1.358
CH3(CH2)3CH3
969132
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15g/mol
CHROMASOLV™
Videos
Safety and Handling

Safety and Handling

P210-P261-P273-P301 + P310-P331

H225-H304-H336-H411

EINECSNumber : 203-692-4

RTECSNumber : RZ9450000

SDS
missing translation for 'documents'

missing translation for 'documents'