missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, AMD CHROMASOLV™, ≥99%, Honeywell Riedel-de Haën™
CHROMASOLV™ AMD,≥99%
$559.10 - $559.10
Chemical Identifiers
CAS | 109-66-0 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009498 |
InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
PubChem CID | 8003 |
ChEBI | CHEBI:37830 |
IUPAC Name | pentane |
SMILES | CCCCC |
Description
- Solvents designed for high-performance thin-layer chromatography
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CHEBI:37830 | |
CCCCC |
Specifications
-130°C (literature) | |
≤100.0000% | |
36°C to 37°C | |
<−35°C | |
C5H12 | |
MFCD00009498 | |
969132 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15 | |
High Purity |
109-66-0 | |
0.626Â g/mLÂ at 25Â °C(lit.) | |
2.5 L | |
Glass Bottle | |
CH3(CH2)3CH3 | |
UN1265 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CCCCC | |
72.15 | |
CHEBI:37830 | |
>99% | |
Pentane, AMD CHROMASOLV™, ≥99% |
Safety and Handling
ShelfLife : 1800 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Pentanes
EINECSNumber : 203-692-4
Recommended Storage : Room Temp