missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentamethylene Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) 97.0+%, TCI America™
[Reagent for application of the exciton chirality method]
$274.02 - $1593.08
Chemical Identifiers
CAS | 210769-64-5 |
---|---|
Molecular Formula | C95H64N8O4Zn2 |
Molecular Weight (g/mol) | 1512.369 |
MDL Number | MFCD02093489 |
InChI Key | NBWTUFLOLJEGJZ-UHFFFAOYSA-L |
Synonym | 1,5-Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoyloxy]pentane Dizinc(II), Pentamethylene Bis[4-(10,15,20-triphenylporphin-5-yl)benzoate]dizinc(II) |
PubChem CID | 53384584 |
SMILES | C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=C(C=C8)C(=O)OCCCCCOC(=O)C9=CC=C(C=C9)C1=C2C=CC(=N2)C(=C2C=CC(=C(C5=NC(=C(C6=CC=C1[N-]6)C1=CC=CC=C1)C=C5)C1=CC=CC=C1)[N-]2)C1=CC=CC=C1)C=C4)C1=CC=CC=C1)[N-]3.[Zn+2].[Zn+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
P136410MG
|
TCI America
P136410MG |
10 mg |
Each for $274.02
|
|
|||||
P1364100MG
|
TCI America
P1364100MG |
100 mg |
Each for $1,593.08
|
|
|||||
Chemical Identifiers
210769-64-5 | |
1512.369 | |
NBWTUFLOLJEGJZ-UHFFFAOYSA-L | |
53384584 |
C95H64N8O4Zn2 | |
MFCD02093489 | |
1,5-Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoyloxy]pentane Dizinc(II), Pentamethylene Bis[4-(10,15,20-triphenylporphin-5-yl)benzoate]dizinc(II) | |
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=C(C=C8)C(=O)OCCCCCOC(=O)C9=CC=C(C=C9)C1=C2C=CC(=N2)C(=C2C=CC(=C(C5=NC(=C(C6=CC=C1[N-]6)C1=CC=CC=C1)C=C5)C1=CC=CC=C1)[N-]2)C1=CC=CC=C1)C=C4)C1=CC=CC=C1)[N-]3.[Zn+2].[Zn+2] |
Specifications
210769-64-5 | |
C95H64N8O4Zn2 | |
10 mg | |
NBWTUFLOLJEGJZ-UHFFFAOYSA-L | |
1512.369 | |
1512.37 | |
Crystalline Powder |
Purple | |
MFCD02093489 | |
1,5-Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoyloxy]pentane Dizinc(II), Pentamethylene Bis[4-(10,15,20-triphenylporphin-5-yl)benzoate]dizinc(II) | |
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=C(C=C8)C(=O)OCCCCCOC(=O)C9=CC=C(C=C9)C1=C2C=CC(=N2)C(=C2C=CC(=C(C5=NC(=C(C6=CC=C1[N-]6)C1=CC=CC=C1)C=C5)C1=CC=CC=C1)[N-]2)C1=CC=CC=C1)C=C4)C1=CC=CC=C1)[N-]3.[Zn+2].[Zn+2] | |
53384584 | |
≥97.0% (HPLC) | |
Pentamethylene Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) [Reagent for application |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator