missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylcyclopentadienylhafnium(IV) trichloride, 98+%

CAS: 75181-08-7 | C10H20Cl3Hf-9 | 425.11 g/mol

$126.16 - $477.84

Chemical Identifiers

CAS 75181-08-7
Molecular Formula C10H20Cl3Hf-9
Molecular Weight (g/mol) 425.11
MDL Number MFCD00070461
InChI Key DROXWYIBBCYOSR-UHFFFAOYSA-K
Synonym pentamethylcyclopentadienylhafnium trichloride, acmc-20ajym, hafnium,trichloro 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
PubChem CID 53471594
IUPAC Name carbanide;cyclopenta-1,3-diene;hafnium;trichloride
SMILES [CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Cl-].[Cl-].[Cl-].[Hf]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2701003
View Documents
Thermo Scientific Chemicals
H2701003
1 g
Each for $126.16
Only null left
 
AAH2701006
View Documents
Thermo Scientific Chemicals
H2701006
5 g
Each for $477.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75181-08-7
425.11
DROXWYIBBCYOSR-UHFFFAOYSA-K
53471594
[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Cl-].[Cl-].[Cl-].[Hf]
C10H20Cl3Hf-9
MFCD00070461
pentamethylcyclopentadienylhafnium trichloride, acmc-20ajym, hafnium,trichloro 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
carbanide;cyclopenta-1,3-diene;hafnium;trichloride

Specifications

75181-08-7
C10H20Cl3Hf-9
1 g
Moisture sensitive
DROXWYIBBCYOSR-UHFFFAOYSA-K
carbanide;cyclopenta-1,3-diene;hafnium;trichloride
53471594
≥98%
170°C to 172°C
MFCD00070461
UN3261
pentamethylcyclopentadienylhafnium trichloride, acmc-20ajym, hafnium,trichloro 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Cl-].[Cl-].[Cl-].[Hf]
425.11
420.08
Pentamethylcyclopentadienylhafnium(IV) trichloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only