Learn More
Gibco™ PBS, pH 7.4
Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order.
Supplier:  Gibco™ A58313BD
 View more versions of this product
Catalog No. A58313BD
Only null left
Explore available promotions
Description
Description
Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order. Commonly used for processing cells and flushing equipment, sourcing Gibco balanced salt solutions can help streamline your operation by reducing buffer preparation and allowing you to focus on additional critical steps in your biopharmaceutical processes. Discover new outsourcing efficiencies to reclaim time, space, and resources.
Product Features
  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification
Benefits
  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals
Specifications
Specifications
None
Phosphate Buffered Saline (PBS)
280 to 315 mOsm/kg
Sterile-filtered
Drum
Clear, colorless solution
50 L
7.3 to 7.5
Liquid
cGMP
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.