missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palonosetron Hydrochloride 98.0+%, TCI America™
$366.62 - $1260.48
Chemical Identifiers
CAS | 135729-62-3 |
---|---|
Molecular Formula | C19H25ClN2O |
Molecular Weight (g/mol) | 332.87 |
MDL Number | MFCD00939524 |
InChI Key | OLDRWYVIKMSFFB-KPVRICSOSA-N |
PubChem CID | 6918303 |
ChEBI | CHEBI:85157 |
IUPAC Name | hydrogen (5R)-3-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3-azatricyclo[7.3.1.0âµ,¹³]trideca-1(13),9,11-trien-2-one chloride |
SMILES | [H+].[Cl-].O=C1N(C[C@@H]2CCCC3=CC=CC1=C23)[C@@H]1CN2CCC1CC2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
P2051200MG
|
TCI America
P2051200MG |
200 mg |
Each for $366.62
|
|
|||||
P20511G
|
TCI America
P20511G |
1 g |
Each for $1,260.48
|
|
|||||
Chemical Identifiers
135729-62-3 | |
332.87 | |
OLDRWYVIKMSFFB-KPVRICSOSA-N | |
CHEBI:85157 | |
[H+].[Cl-].O=C1N(C[C@@H]2CCCC3=CC=CC1=C23)[C@@H]1CN2CCC1CC2 |
C19H25ClN2O | |
MFCD00939524 | |
6918303 | |
hydrogen (5R)-3-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3-azatricyclo[7.3.1.0âµ,¹³]trideca-1(13),9,11-trien-2-one chloride |
Specifications
135729-62-3 | |
C19H25ClN2O | |
200 mg | |
[H+].[Cl-].O=C1N(C[C@@H]2CCCC3=CC=CC1=C23)[C@@H]1CN2CCC1CC2 | |
332.87 | |
CHEBI:85157 | |
≥98.0% (HPLC,T) | |
Palonosetron Hydrochloride |
307°C | |
MFCD00939524 | |
OLDRWYVIKMSFFB-KPVRICSOSA-N | |
hydrogen (5R)-3-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3-azatricyclo[7.3.1.0âµ,¹³]trideca-1(13),9,11-trien-2-one chloride | |
6918303 | |
332.87 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator