Learn More
Palmityl palmitate, 96%
CAS: 540-10-3 | C32H64O2 | 480.86 g/mol
$154.47 - $517.23
Chemical Identifiers
CAS | 540-10-3 |
---|---|
Molecular Formula | C32H64O2 |
Molecular Weight (g/mol) | 480.86 |
MDL Number | MFCD00053739 |
InChI Key | PXDJXZJSCPSGGI-UHFFFAOYSA-N |
Synonym | cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester |
PubChem CID | 10889 |
ChEBI | CHEBI:75584 |
IUPAC Name | hexadecyl hexadecanoate |
SMILES | CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5928303
|
Thermo Scientific Chemicals
H5928303 |
1 g |
Each for $154.47
|
|
|||||
AAH5928306
|
Thermo Scientific Chemicals
H5928306 |
5 g |
Each for $517.23
|
|
|||||
Description
It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.
Solubility
Soluble in hot acetone.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
Chemical Identifiers
540-10-3 | |
480.86 | |
PXDJXZJSCPSGGI-UHFFFAOYSA-N | |
10889 | |
hexadecyl hexadecanoate |
C32H64O2 | |
MFCD00053739 | |
cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester | |
CHEBI:75584 | |
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC |
Specifications
540-10-3 | |
C32H64O2 | |
MFCD00053739 | |
cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester | |
PXDJXZJSCPSGGI-UHFFFAOYSA-N | |
hexadecyl hexadecanoate | |
10889 | |
480.85 | |
Palmityl palmitate |
55°C to 56°C | |
CH3(CH2)14CO2(CH2)15CH | |
1 g | |
Soluble in hot acetone. | |
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC | |
480.86 | |
CHEBI:75584 | |
96% |
Safety and Handling
EINECSNumber : 208-736-6
RTECSNumber : RT4750000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only