missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic Acid Monohydrate 98.0+%, TCI America™

$39.14 - $55.92

Chemical Identifiers

CAS 6192-52-5
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
MDL Number MFCD00064387
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
PubChem CID 521998
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T026725G
View Documents
TCI America
T026725G
25 g
Each for $39.14
Only null left
 
T0267500G
View Documents
TCI America
T0267500G
500 g
Each for $55.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6192-52-5
190.213
KJIFKLIQANRMOU-UHFFFAOYSA-N
521998
CC1=CC=C(C=C1)S(=O)(=O)O.O
C7H10O4S
MFCD00064387
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
4-methylbenzenesulfonic acid;hydrate

Specifications

106°C
140°C
C7H10O4S
2585
KJIFKLIQANRMOU-UHFFFAOYSA-N
4-methylbenzenesulfonic acid;hydrate
521998
≥98.0% (HPLC,T)
Crystalline Powder
6192-52-5
White
MFCD00064387
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
CC1=CC=C(C=C1)S(=O)(=O)O.O
190.213
172.20
25 g
p-Toluenesulfonic Acid Monohydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1901

RTECSNumber : DB7164000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'