missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxcarbazepine 98.0+%, TCI America™
$167.76 - $458.56
Chemical Identifiers
CAS | 28721-07-5 |
---|---|
Molecular Formula | C15H12N2O2 |
Molecular Weight (g/mol) | 252.273 |
MDL Number | MFCD00865307 |
InChI Key | CTRLABGOLIVAIY-UHFFFAOYSA-N |
Synonym | oxcarbazepine, trileptal, oxcarbamazepine, oxacarbazepine, oxcarbazepina, oxcarbazepinum, timox, oxcarbazepinum inn-latin, oxcarbazepina inn-spanish, oxcarbazepine inn |
PubChem CID | 34312 |
ChEBI | CHEBI:7824 |
IUPAC Name | 5-oxo-6H-benzo[b][1]benzazepine-11-carboxamide |
SMILES | C1C2=CC=CC=C2N(C3=CC=CC=C3C1=O)C(=O)N |
Chemical Identifiers
28721-07-5 | |
252.273 | |
CTRLABGOLIVAIY-UHFFFAOYSA-N | |
34312 | |
5-oxo-6H-benzo[b][1]benzazepine-11-carboxamide |
C15H12N2O2 | |
MFCD00865307 | |
oxcarbazepine, trileptal, oxcarbamazepine, oxacarbazepine, oxcarbazepina, oxcarbazepinum, timox, oxcarbazepinum inn-latin, oxcarbazepina inn-spanish, oxcarbazepine inn | |
CHEBI:7824 | |
C1C2=CC=CC=C2N(C3=CC=CC=C3C1=O)C(=O)N |
Specifications
28721-07-5 | |
C15H12N2O2 | |
5 g | |
CTRLABGOLIVAIY-UHFFFAOYSA-N | |
5-oxo-6H-benzo[b][1]benzazepine-11-carboxamide | |
34312 | |
252.27 | |
Crystalline Powder |
216°C | |
MFCD00865307 | |
oxcarbazepine, trileptal, oxcarbamazepine, oxacarbazepine, oxcarbazepina, oxcarbazepinum, timox, oxcarbazepinum inn-latin, oxcarbazepina inn-spanish, oxcarbazepine inn | |
C1C2=CC=CC=C2N(C3=CC=CC=C3C1=O)C(=O)N | |
252.273 | |
CHEBI:7824 | |
≥98.0% (HPLC,N) | |
Oxcarbazepine |
Safety and Handling
RTECSNumber : HN8445000
TSCA : No