missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid, Dihydrate, Crystal, ACS, 99.5-102.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor O102525KG
Specifications
6153-56-6 | |
C2H6O6 | |
O.O.OC(=O)C(O)=O | |
126.06 | |
ACS | |
Poly Bottle |
1 | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
oxalic acid dihydrate | |
99.5 to 102.5% | |
2.5 kg | |
0.0001 |
Chemical Identifiers
6153-56-6 | |
126.06 | |
oxalic acid dihydrate |
C2H6O6 | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
O.O.OC(=O)C(O)=O |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.