missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, ACS Reagent, Reagents

Oxalic Acid Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

$559.50 - $5278.70

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.06
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym Ethanedioic Acid dihydrate
IUPAC Name oxalic acid dihydrate
SMILES O.O.OC(=O)C(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-338-168
View Documents
Labchem Inc
C111050012A5
12 kg
Each for $1,750.47
Only null left
 
01-338-167
View Documents
Labchem Inc
C111050025F4
25 kg
Each for $2,872.50
Only null left
 
01-338-166
View Documents
Labchem Inc
C1110500500A5
500 g
Each for $559.50
Only null left
 
01-338-165
View Documents
Labchem Inc
C111050050KG
50 kg
Each for $5,278.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6153-56-6
126.06
Ethanedioic Acid dihydrate
O.O.OC(=O)C(O)=O
C2H6O6
GEVPUGOOGXGPIO-UHFFFAOYSA-N
oxalic acid dihydrate

Specifications

101°C to 102°C
Colorless/White
Ethanedioic Acid dihydrate
O.O.OC(=O)C(O)=O
126.06
12 kg
Solid
Oxalic Acid Dihydrate, ACS Reagent
6153-56-6
C2H6O6
GEVPUGOOGXGPIO-UHFFFAOYSA-N
oxalic acid dihydrate
ACS
Poly Pail
1.653
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'