missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite Outdoor Flammable Cabinet
Engineered to withstand the elements and potential impacts, this cabinet is OSHA, NFPA, and FM approved, ensuring your flammable chemicals are safely and securely stored outdoors.
Supplier: Justrite 814500
Description
- Spill Slope shelves with fully welded sump
- Ground Screw
- Durable and Chemical - resistant powder coated paint
- UV resistant
- Vent with built in flash arrestors
- Welded Shelf Hangers
- Leak Proof Sump
Specifications
45 gal. | |
Safe outdoor storage of flammable liquids to avoid risk of potential fire hazards and chemical spills in industries like chemical manufacturing, petrochemical and laboratories | |
18 x 43 x 65 in. | |
14.6 x 39.5 x 60 in. | |
Adjustable | |
Manual-close, Hinged, Three-point lock, Locking | |
2 |
Flammables Storage Cabinet | |
Steel (18 ga) | |
120 kg | |
Yellow | |
264.7 lb. | |
2 | |
OSHA, NFPA, and FM approved |