missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 500 ppm C, Ricca Chemical

$278.69 - $437.28

Chemical Identifiers

CAS 7732-18-5
Molecular Formula C8H5KO4
Molecular Weight (g/mol) Mixture
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R1846500250
View Documents
Ricca Chemical Company
R1846500250C
250 mL Amber Glass Bottle
Each for $278.69
Only null left
 
R18465001C
View Documents
Ricca Chemical Company
R18465001C
1 L Amber Glass Bottle
Each for $437.28
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
Mixture
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate

Specifications

Amber Glass Bottle
Odorless
C8H5KO4
100°C
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
250 mL
Laboratory
7732-18-5,7664-38-2,877-24-7
Liquid
Colorless
Standard
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Mixture
23676735
Organic Carbon Standard, 500 ppm C
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS
missing translation for 'documents'

missing translation for 'documents'