missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ One-Hour Fire Barrier

Justrite™ One-Hour Fire Barrier is for applications where both fuels and oxidizers are stored in close proximity

$1991.77 - $1991.77

Specifications

Type One-Hour Fire Barrier
Material Steel, Marinite I
Color Gray
Finish Powder-coated Paint
For Use With (Equipment) Gas cylinders
View More Specs

Products 1
Catalog Number Mfr. No. Holds Price Quantity  
Catalog Number Mfr. No. Holds Price Quantity  
19-085-848
View Documents
Justrite™
35352
1 Cylinder
Each for $1,991.77
Only null left
 
Description

Description

  • Barrier between barricade racks, mounting brackets, or stands exceeds minimum NFPA 55, OSHA and CGA standards for separation of oxidizers and flammable gases
  • Constructed using fire-resistant Marinite™ I
  • Fully encased into welded steel panels
  • May be mounted to exterior walls and floor using provided mounting holes with self-supplied fasteners, or be used as a free-standing wall
  • Lift hook plates supplied for installation positioning

Compliance

OSHA, CGA P1, NFPA55

Warranty

1 YEAR

Specifications

Specifications

One-Hour Fire Barrier
Gray
Gas cylinders
Steel, Marinite I
Powder-coated Paint
51 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'