missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleanolic Acid Hydrate 98.0+%, TCI America™
$141.67 - $438.05
Chemical Identifiers
CAS | 508-02-1 |
---|---|
Molecular Formula | C30H48O3 |
Molecular Weight (g/mol) | 456.711 |
MDL Number | MFCD00064914 |
InChI Key | MIJYXULNPSFWEK-GTOFXWBISA-N |
Synonym | oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid |
PubChem CID | 10494 |
ChEBI | CHEBI:37659 |
IUPAC Name | (4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid |
SMILES | CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C |
Chemical Identifiers
508-02-1 | |
456.711 | |
MIJYXULNPSFWEK-GTOFXWBISA-N | |
10494 | |
(4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid |
C30H48O3 | |
MFCD00064914 | |
oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid | |
CHEBI:37659 | |
CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C |
Specifications
508-02-1 | |
White | |
MFCD00064914 | |
oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid | |
CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C | |
456.711 | |
CHEBI:37659 | |
≥98.0% (T) | |
Oleanolic Acid Hydrate |
311°C | |
C30H48O3 | |
5 g | |
MIJYXULNPSFWEK-GTOFXWBISA-N | |
(4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid | |
10494 | |
456.71 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : RK0177965
TSCA : Yes