missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Ocean Pacific Ninja ™ Nitrile Gloves

Fully textured, powder free black nitrile glove with superior resistance and grip. Puncture resistance provides enhanced protection against bloodborne pathogens, chemical lubricants and sharp instruments.

$22.32 - $22.32

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Black
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
296367492
View Documents
Medicom™
MEDOPSNI
Small
Pack of 200 Each for $22.32
Case of 10 Pack for $223.20
Only null left
 
296367493
View Documents
Medicom™
MEDOPMNI
Medium
Pack of 200 Each for $22.32
Case of 10 Pack for $223.20
Only null left
 
Description

Description

  • Thick and durable
  • Fully textured to ensure a secure grip, even when wet
  • Punture resistant
  • Powder free
  • Industries include tattoo, hair design, dentistry, ambulance, laboratory science or pet care
  • ASTM D6978-05 certified for chemotherapy handling and CFIA certified for food handling
Specifications

Specifications

No
Healthcare, Industrial, Laboratory
Black
Fully Textured
Powder Free
Yes
No
1.5
Single-use
Yes
Non-sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'