missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Oakton™ Universal AC Power Adapters
Oakton Universal AC Power Adapters work globally and come with multiple wall outlet adapters.
Supplier: Cole-Parmer™ 3561507
Description
9 Volt, 6 Watt to 100/240 VAC, 50/60 HzSpecifications
Black | |
4.33 x 2.75 in. | |
4.33 x 2.75 x 2.25 in. | |
Conductivity, DO, pH | |
AC adapter | |
10.998 cm | |
0.4 lb. | |
0.18 kg | |
6.985 cm |
Oakton Universal AC Adapter for 510/550/560/700/2100 Series | |
Accessory Power Adapter | |
9 Volt, 6 Watt, 100/240 VAC, 50/60 Hz | |
50/60 Hz | |
4.33 in. | |
100/240 V | |
6 Months | |
2.75 in. |