missing translation for 'onlineSavingsMsg'
Learn More

o-Phthalaldehyde, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P113925GM

 View more versions of this product

Catalog No. 18-612-084


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

643-79-8
53° to 58°C
Amber Glass Bottle
25 g
O=CC1=CC=CC=C1C=O
134.13
Crystals or Flakes
1
83°C
C8H6O2
ZWLUXSQADUDCSB-UHFFFAOYSA-N
benzene-1,2-dicarbaldehyde
Reagent

Chemical Identifiers

643-79-8
134.13
benzene-1,2-dicarbaldehyde
C8H6O2
ZWLUXSQADUDCSB-UHFFFAOYSA-N
O=CC1=CC=CC=C1C=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.