missing translation for 'onlineSavingsMsg'
Learn More

O-Ethylhydroxylamine hydrochloride, 99.0-101.0 wt. % (AT), MilliporeSigma™ Supelco™

$775.19 - $775.19

Chemical Identifiers

MDL Number MFCD00012956
Synonym Ethoxyamine hydrochloride
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111013947
View Documents
Merck Emd Millipore
4350410G
10 g
Each for $775.19
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00012956
Ethoxyamine hydrochloride

Specifications

130°C to 133°C
CH3 CH2 ONH2 · HCl
10 g
97.54
For GC derivatization
3332-29-4
MFCD00012956
Ethoxyamine hydrochloride
99.0 to 101.0 wt. % (AT)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'