missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech NuTech™ Lab Coats
Lightweight and breathable microporous composite material offers comfort and protection for a broad range of non-hazardous applications.
$194.20 - $237.35
Specifications
Product Type | Controlled Environments Lab Coat |
---|---|
Material | Microporous Composite Fabric |
Color | White |
Closure Type | Snaps |
Cuff Style | Elastic |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-810-573
|
AlphaProTech
LC626212 |
Small/Medium |
Case of 25 Each for $194.20
|
|
|||||
19-810-574
|
AlphaProTech
LC626214 |
Large/X-Large |
Case of 25 Each for $194.20
|
|
|||||
19-810-575
|
AlphaProTech
LC626216 |
2X-Large/3X-Large |
Case of 25 Each for $214.44
|
|
|||||
19-810-576
|
AlphaProTech
LC626218 |
4X-Large/5X-Large |
Case of 25 Each for $237.35
|
|
|||||
Description
- Strong breathable material offering protection
- Non-confining tapered collar designed like shirt collar for style and comfort
- Five-snap closure to provide fill coverage without gaps
- Length approximately adjusted to size to provide additional coverage
- Features tapered collar, elastic wrists, 3 pockets, snap close and serged seams
Specifications
Controlled Environments Lab Coat | |
White | |
Elastic | |
3 |
Microporous Composite Fabric | |
Snaps | |
Single-use | |
Serged |