missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm DFS Item

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system.

$398.00 - $415.00

Specifications

Gel Percentage 3 to 8%
Gel Size Midi
GelThickness 1.0 mm
Gel Type Tris-Acetate
Recommended Applications Denaturing, Native
View More Specs

Products 6
Catalog Number Mfr. No. Wells Price Quantity  
Catalog Number Mfr. No. Wells Price Quantity  
WG1601BOX DFS Item
View Documents
Invitrogen™
WG1601BOX
12 + 2-well
Each for $398.00
Only null left
 
WG1601A DFS Item
View Documents
Invitrogen™
WG1601A
12 + 2-well, w/adapters
Each for $415.00
Only null left
 
WG1602BOX DFS Item
View Documents
Invitrogen™
WG1602BOX
20-well
Each for $398.00
Only null left
 
WG1602A DFS Item
View Documents
Invitrogen™
WG1602A
20-well, w/adapters
Each for $412.00
Only null left
 
WG1603BOX DFS Item
View Documents
Invitrogen™
WG1603BOX
26-well
Each for $401.00
Only null left
 
WG1603A DFS Item
View Documents
Invitrogen™
WG1603A
26-well, w/adapters
Each for $415.00
Only null left
 
Description

Description

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of high molecular weight proteins compared to traditional Tris-glycine SDS-PAGE gels.

Features of NuPAGE Tris-Acetate gels:
High resolution—gels offer optimal separation of high molecular weight proteins
Better protein integrity—sample preparation process has been optimized to help preserve proteins
Longer self life—gels can be stored for at least eight months

Choose the right NuPAGE Tris-Acetate gel for your protein separation
Obtain optimal separation of your high molecular weight proteins by choosing the right combination of gel and running buffer. NuPAGE Tris-Acetate protein gels come in a polyacrylamide concentration of 7% and a 3–8% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Tris-Acetate gels also come in multiple well formats. Mini gels can be run using our XCell SureLock Mini-Cell (EI0001) or Mini Gel Tank (A25977). Midi gels can be run using our XCell4 SureLock Midi-Cell (WR0100) or conveniently with the Bio-Rad Criterion™ Cell using our adapters (WA0999).

Run your proteins in native or denatured form
NuPAGE Tris-Acetate protein gels do not contain SDS and can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Tris-Acetate SDS Running Buffer (LA0041). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC26720.

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP00061) for traditional wet transfer using the XCell II Blot Module (EI9051) or the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB210010).

Specifications

Specifications

3 to 8%
1.0 mm
Denaturing, Native
Denaturing, Native
NuPAGE™
Wet Ice
8 cm
Midi
Tris-Acetate
36 to 500 kDa
Molecular Weight
10 gels (1 box)
Store at 2°C to 8°C. Do not freeze.
13 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'