missing translation for 'onlineSavingsMsg'
Learn More

Norlaudanosine Hydrochloride 98.0+%, TCI America™

$76.43 - $307.57

Chemical Identifiers

CAS 6429-04-5
Molecular Formula C20H26ClNO4
Molecular Weight (g/mol) 379.88
MDL Number MFCD00035267
InChI Key VMPLLPIDRGXFTQ-UHFFFAOYNA-N
Synonym Tetrahydropapaverine Hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride
PubChem CID 16667431
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES COC1=C(C=C(C=C1)CC2C3=CC(=C(C=C3CCN2)OC)OC)OC.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09185G
View Documents
TCI America
N09185G
5 g
Each for $76.43
Only null left
 
N091825G
View Documents
TCI America
N091825G
25 g
Each for $307.57
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6429-04-5
379.88
VMPLLPIDRGXFTQ-UHFFFAOYNA-N
16667431
COC1=C(C=C(C=C1)CC2C3=CC(=C(C=C3CCN2)OC)OC)OC.Cl
C20H26ClNO4
MFCD00035267
Tetrahydropapaverine Hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride
1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride

Specifications

6429-04-5
White
MFCD00035267
Tetrahydropapaverine Hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride
COC1=C(C=C(C=C1)CC2C3=CC(=C(C=C3CCN2)OC)OC)OC.Cl
379.88
379.88
Crystalline Powder
218°C
C20H26ClNO4
5 g
VMPLLPIDRGXFTQ-UHFFFAOYNA-N
1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
16667431
≥98.0% (HPLC,N)
Norlaudanosine Hydrochloride
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NX5070700

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'