missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Non Metals, plasma standard solution, Specpure™, As, B, P, S, Se, Si, Te at 100μg/mL
$646.10 - $646.10
Specifications
Chemical Name or Material | Non Metals plasma standard solution |
---|---|
Name Note | As, B, P, S, Se, Si, Te at 100μg/mL |
Physical Form | Liquid |
MDL Number | MFCD00144997 |
UN Number | UN3264 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Non Metals plasma standard solution | |
Liquid | |
UN3264 |
As, B, P, S, Se, Si, Te at 100μg/mL | |
MFCD00144997 | |
Specpure™ |
RUO – Research Use Only