missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane, 96%, for HPLC

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol

$62.31 - $494.62

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
Synonym methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
PubChem CID 6375
ChEBI CHEBI:77701
IUPAC Name nitromethane
SMILES C[N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445671000
SDS View Documents
Thermo Scientific Chemicals
445671000
100 mL N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445670010
SDS View Documents
Thermo Scientific Chemicals
445670010
1 L N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445670025
SDS View Documents
Thermo Scientific Chemicals
445670025
2.5 L N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-52-5
61.04
LYGJENNIWJXYER-UHFFFAOYSA-N
6375
nitromethane
CH3NO2
MFCD00007400
methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
CHEBI:77701
C[N+]([O-])=O

Specifications

75-52-5
1.1200g/mL
35°C
Glass bottle
1.3805 to 1.3825
MFCD00007400
1.12
methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
LYGJENNIWJXYER-UHFFFAOYSA-N
nitromethane
6375
CHEBI:77701
96%
Nitromethane
-29.0°C
100.0°C to 102.0°C
95% min. (GC)
CH3NO2
CH3NO2
100 mL
15,6698
Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide
C[N+]([O-])=O
61.04
0.61 mPa.s (25°C)
61.04
HPLC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh

GHS Signal Word: Warning

EINECSNumber : 200-876-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only