missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrobenzene, ACS Reagent, Reagents
Nitrobenzene, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$654.10 - $1,962.30
Chemical Identifiers
CAS | 98-95-3 |
---|---|
Molecular Formula | C6H5NO2 |
Molecular Weight (g/mol) | 123.11 |
InChI Key | LQNUZADURLCDLV-UHFFFAOYSA-N |
Synonym | Nitrobenzol, Oil of Mirbane |
IUPAC Name | nitrobenzene |
SMILES | [O-][N+](=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-337-881
|
Labchem Inc
C12575001C |
1 L |
N/A
|
|
|||||
01-337-880
|
Labchem Inc
C12575004C |
4 L |
N/A
|
|
|||||
Chemical Identifiers
98-95-3 | |
123.11 | |
Nitrobenzol, Oil of Mirbane | |
[O-][N+](=O)C1=CC=CC=C1 |
C6H5NO2 | |
LQNUZADURLCDLV-UHFFFAOYSA-N | |
nitrobenzene |
Specifications
98-95-3 | |
Pale Yellow | |
Glass Bottle | |
1 L | |
Nitrobenzol, Oil of Mirbane | |
[O-][N+](=O)C1=CC=CC=C1 | |
123.11 | |
Liquid |
5.6°C to 5.8°C | |
211°C | |
C6H5NO2 | |
1.2037 | |
LQNUZADURLCDLV-UHFFFAOYSA-N | |
nitrobenzene | |
ACS | |
Nitrobenzene, ACS Reagent |