missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, TRACESELECT™ Ultra, for UltraTrace Analysis, 67-69%, Honeywell Fluka™

TraceSELECT™ Ultra, for ultratrace analysis, 67-69%

$1089.70 - $5770.73

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044473
View Documents
Honeywell Chemicals
02650250ML
250 mL Poly Bottle
Each for $1,089.70
Only null left
 
6001110
View Documents
Honeywell Chemicals
02650500ML
500 mL Poly bottle
Each for $1,654.88
Only null left
 
60044474
View Documents
Honeywell-Fluka
026502L
2 L Poly Bottle
Each for $5,770.73
Only null left
 
Description

Description

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Specifications

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O

Specifications

7697-37-2
1.420 g/cm3
8mmHg (20°C)
250 mL
HNO3
UN2031
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
63.01
Ultratrace™
122°C
Poly Bottle
64.5 to 70%
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
O[N+]([O-])=O
63.01
CHEBI:48107
67 to 69%
Nitric acid, TRACESELECT™ Ultra, for UltraTrace Analysis, 65-71%,
Videos
Safety and Handling

Safety and Handling

P210-P220-P260-P280-P305 + P351 + P338-P370 + P378

H272-H290-H314

SDS
missing translation for 'documents'

missing translation for 'documents'