missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 0.250 N (N/4), Reagents
Nitric Acid, 0.250 N (N/4), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Labchem Inc CN2065004A
Specifications
0.6 | |
Colorless | |
100°C | |
1.0085 | |
H2O | |
Azotic Acid, Hydrogen nitrate | |
O | |
18.02 |
Nitric Acid, 0.250 N (N/4) | |
7732-18-5,7697-37-2 | |
Poly Bottle | |
4 L | |
Liquid | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent |
Chemical Identifiers
7732-18-5 | |
18.02 | |
Azotic Acid, Hydrogen nitrate | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |