missing translation for 'onlineSavingsMsg'
Learn More

Niobium foil, 0.3mm (0.01in) thick, annealed, 99.8% (metals basis)

CAS: 7440-03-1 | Nb | 92.91 g/mol

$103.79 - $1082.14

Chemical Identifiers

CAS 7440-03-1
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40668FY
View Documents
Thermo Scientific Chemicals
040668FY
50 x 100 mm
Each for $103.79
Only null left
 
AA40668GK
View Documents
Thermo Scientific Chemicals
040668GK
100 x 200 mm
Each for $278.92
Only null left
 
AA40668HQ
View Documents
Thermo Scientific Chemicals
040668HQ
200 x 500 mm
Each for $1,082.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
CHEBI:33344
[Nb]

Specifications

7440-03-1
Odorless
Foil
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
23936
99.8%
(metals basis)
50 x 100 mm
≈12.9g/50x100mm
Nb
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
[Nb]
92.91
CHEBI:33344
Niobium foil, 0.3mm (0.01 in.) thick, Annealed
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-113-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only