missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$163.38 - $372.00
Chemical Identifiers
CAS | 7440-02-0 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 MFCD06798735 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ni | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
CHEBI:28112 | |
[Ni] |
Ni | |
MFCD00011137 MFCD06798735 | |
935 | |
nickel |
Specifications
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
0.1,2.0,97.9 | |
MFCD00011137 MFCD06798735 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
ICP/ICP-MS Standard | |
[Ni] | |
nickel | |
30 mL | |
CHEBI:28112 | |
Nickel Standard |
7440-02-0,7697-37-2,7732-18-5 | |
0.1,2.0,97.9 | |
Ni | |
ICP-MS | |
Standard Solution | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
2% HNO3 | |
58.69 | |
935 | |
ICP/ICP-MS |
Safety and Handling
Causes severe skin burns and eye damage
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only